missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 7 Mouse anti-Human, Clone: 2E1G5, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66483-1-IG-20UL

 View more versions of this product

Product Code. 16808405

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specifically against KRT7.

Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
Specifications

Specifications

Cytokeratin 7
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08729
KRT7
Cytokeratin 7 Fusion Protein ag7895
20 μL
Primary
Human
Antibody
IgG2a
Western Blot, Immunohistochemistry (Paraffin)
2E1G5
Unconjugated
KRT7
CK 7, CK7, Cytokeratin 7, K2C7, K7, keratin 7, KRT7, Sarcolectin, SCL, Type II keratin Kb7
Mouse
Protein A
RUO
3855
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.