Learn More
Invitrogen™ Cytokeratin 7 Monoclonal Antibody (RCK105)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA106315
Description
MA1-06315 detects cytokeratin 7 in human, mouse, rat, hamster, canine, feline, goat and zebrafish samples. MA1-06315 has sucessfully been used in Western blotting, flow cytometry, immunocytochemistry and immunohistochemistry. The MA1-06315 immunogens are cytokeratins from the human bladder carcinoma cell line T24. Store at 4ºC or in small aliquots at -20ºC.
Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
Specifications
| Cytokeratin 7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P08729, Q6IG12, Q9DCV7 | |
| KRT7, LOC100755511, LOC100861181, si:ch73-233m11.2 | |
| Cytokeratins from the human bladder carcinoma cell line T24. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Hamster, Feline, Zebrafish, Goat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| RCK105 | |
| Unconjugated | |
| KRT7 | |
| CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; NACHT, LRR and PYD domains-containing protein 3; sarcolectin; SCL; si:ch73-233m11.1; si:ch73-233m11.2; similar to keratin 7; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729} | |
| Mouse | |
| Purified | |
| RUO | |
| 100331661, 100755511, 100861181, 101096243, 102184596, 110310, 300242, 3855, 477602 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.