missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 7 Monoclonal Antibody (OV-TL 12/30)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA511986
This item is not returnable.
View return policy
Description
MA5-11986 targets Cytokeratin 7 in IHC (P) applications and shows reactivity with Human samples. The MA5-11986 immunogen is oTN II ovarian carcinoma cell line.
Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
Specifications
| Cytokeratin 7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| P08729 | |
| KRT7 | |
| OTN II ovarian carcinoma cell line. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OV-TL 12/30 | |
| Unconjugated | |
| KRT7 | |
| CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; NACHT, LRR and PYD domains-containing protein 3; sarcolectin; SCL; si:ch73-233m11.1; si:ch73-233m11.2; similar to keratin 7; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729} | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3855 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction