Learn More
Cytokeratin 7 Antibody (OV-TL12/30), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47940PCP
Product Code. 18657838
  • £508.00 / 0.10mL
Description
Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 7
Monoclonal
PerCP
CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7
OTN 11, ovarian carcinoma cell line
0.1 mL
Cell Biology, Cellular Markers
3855
Store at 4°C in the dark.
IgG1 κ
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunofluorescence
OV-TL12/30
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified