missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7 Antibody (KRT7/8121R), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20720IR
This item is not returnable.
View return policy
Description
Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 7 | |
| Monoclonal | |
| DyLight 755 | |
| CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
| Recombinant fragment (around aa1-200) of human Cytokeratin 7 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3855 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| KRT7/8121R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction