missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 7, 8 Antibody (CAM5.2), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £368.00

Specifications

Antigen Cytokeratin 7, 8
Clone CAM5.2
Concentration 0.2 mg/ml
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30288041
View Documents
Novus Biologicals
NBP3-44630-100ug
100 μg
£368.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30296895
View Documents
Novus Biologicals
NBP3-44630-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytokeratin 7, 8 Monoclonal antibody specifically detects Cytokeratin 7, 8 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

Cytokeratin 7, 8
0.2 mg/ml
Monoclonal
Purified
RUO
Human
CK7, CK-7, CK8, CK-8, Cytokeratin-7, Cytokeratin-8, keratin 8, keratin, type II cytoskeletal 7, keratin-7
Human colorectal carcinoma cell line HT29
Primary
Store at 4⪚C.
CAM5.2
Immunohistochemistry
Unconjugated
Mouse
Cellular Signaling
10 mM PBS with 0.05% BSA
3855
IgG2a κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.