missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 6b Antibody (1C4), Novus Biologicals™

Product Code. 18328878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328878 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328878 Supplier Novus Biologicals Supplier No. H00003854M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytokeratin 6b Monoclonal antibody specifically detects Cytokeratin 6b in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 6b
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C4
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH34535
Gene Alias CK6B, CK-6B, cytokeratin 6B, cytokeratin-6B, K6B, keratin 6B, keratin, epidermal, type II, K6B, keratin, type II cytoskeletal 6B, keratin-6B, keratin-like 1 (a type II keratin sequence), KRTL1, PC2, Type-II keratin Kb10
Host Species Mouse
Immunogen KRT6B (AAH34535, 1 a.a. ~ 564 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MASTSTTIRSHSSSRRGFSANSARLPGVSRSGFSSISVSRSRGSGGLGGACGGAGFGSRSLYGLGGSKRISIGGGSCAISGGYGSRAGGSYGFGGAGSGFGFGGGAGIGFGLGGGAGLAGGFGGPGFPVCPPGGIQEVTVNQSLLTPLNLQIDPAIQRVRAEEREQIKTLNNKFASFIDKVRFLEQQNKVLDTKWTLLQEQGTKTVRQNLEPLFEQYINNLRRQLDNIVGERGRLDSELRNMQDLVEDLKNKYEDEINKRTAAENEFVTLKKDVDAAYMNKVELQAKADTLTDEINFLRALYDAELSQMQTHISDTSVVLSMDNNRNLDLDSIIAEVKAQYEEIAQRSRAEAESWYQTKYEELQITAGRHGDDLRNTKQEIAEINRMIQRLRSEIDHVKKQCANLQAAIADAEQRGEMALKDAKNKLEGLEDALQKAKQDLARLLKEYQELMNVKLALDVEIATYRKLLEGEECRLNGEGVGQVNISVVQSTVSSGYGGASGVGSGLGLGGGSSYSYGSGLGVGGGFSSSSGRATGGGLSSVGGGSSTIKYTTTSSSSRKSYKH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3854
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.