missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584483
This item is not returnable.
View return policy
Description
Immunogen sequence: SGRAIGGGLS SVGGGSSTIK YTTTSSSSRK SYKH Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 74%.
Keratins 6 and 16 are expressed in keratinocytes, which are undergoing rapid turnover in the suprabasal region (also known as hyperproliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyperproliferative situations. Epidermal injury results in activation of keratinocytes which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.
Specifications
| Cytokeratin 6A | |
| Polyclonal | |
| Unconjugated | |
| KRT6A | |
| 2310016L08Rik; 60-kDa keratin; CK6A; CK-6A; CK6C; CK-6C; CK6D; CK-6D; CK-6E; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; cytokeratin-6A; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; Hom s 5; K6A; K6a keratin; K6C; K6D; kamp-keratin derived antimicrobial peptide; KDAMP; Ker2; keratin 6A; keratin 6A, , type II; keratin 6A, type II; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, gene 6a; keratin K6; keratin K6h; keratin, epidermal type II, K6A; keratin, type II cytoskeletal 6A; keratin, type II cytoskeletal 6C; Keratin-6A; keratin-6-alpha; keratin-6C; Krt2-6; Krt2-6a; Krt2-6c; Krt6; KRT6A; KRT6C; KRT6D; LOC683313; mK6[a]; MK6a; mK6-alpha; PC3; similar to keratin complex 2, basic, gene 6a; type-II keratin Kb12; type-II keratin Kb6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P02538 | |
| KRT6A | |
| Recombinant protein corresponding to Human KRT6A. Recombinant protein control fragment (Product #RP-103999). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction