missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytokeratin 6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13453257
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13453257 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13453257 Supplier Invitrogen™ Supplier No. PA529134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (89%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Keratins 6 and 16 are expressed in keratinocytes, which are undergoing rapid turnover in the suprabasal region (also known as hyperproliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyperproliferative situations. Epidermal injury results in activation of keratinocytes which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene KRT6A
Gene Accession No. P02538, P04259, P48668, P50446, Q9Z331
Gene Alias 2310016L08Rik; 60-kDa keratin; BB144589; CK6A; CK-6A; CK6B; CK-6B; CK6C; CK-6C; CK6D; CK-6D; CK-6E; cytokeratin 6A; cytokeratin 6B; cytokeratin 6C; cytokeratin 6D; cytokeratin-6A; Cytokeratin-6B; cytokeratin-6C; Cytokeratin-6D; Cytokeratin-6E; Hom s 5; K6A; K6a keratin; K6B; K6C; K6D; K6E; kamp-keratin derived antimicrobial peptide; KDAMP; Ker2; keratin 6A; keratin 6A, , type II; keratin 6A, type II; keratin 6B; keratin 6B, type II; keratin 6C; keratin 6C, type II; keratin 6E; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6b; keratin complex 2, basic, gene 6c; keratin complex 2, gene 6a; keratin K6; keratin K6h; keratin, epidermal type II, K6A; keratin, epidermal, type II, K6B; keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 6B; keratin, type II cytoskeletal 6C; keratin-6A; Keratin-6-alpha; Keratin-6B; keratin-6-beta; keratin-6C; keratin-like 1 (a type II keratin sequence); Krt2-6; Krt2-6a; Krt2-6b; Krt2-6c; Krt6; Krt6a; Krt6b; KRT6C; KRT6D; KRT6E; KRTL1; LOC683313; mK6[a]; mK6[b]; MK6a; mK6-alpha; mK6-beta; PC2; PC3; PC4; PPKNEFD; similar to keratin complex 2, basic, gene 6a; type-II keratin Kb10; Type-II keratin Kb12; type-II keratin Kb6
Gene Symbols KRT6A, KRT6B, KRT6C
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Cytokeratin 6B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16687, 16688, 286887, 3853, 3854
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.