missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 6 Mouse, Clone: LHK6; same as LHK6B, enQuireBio™

Product Code. 15930318
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15930318 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15930318 Supplier EnquireBio Supplier No. 3853MSM1P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen Cytokeratin 6
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Cellular Localization Cytoplasmic
Classification Monoclonal
Clone LHK6; same as LHK6B
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% azide
Gene Accession No. NP_775109
Gene Symbols KRT6A; KRT6B; KRT6C
Host Species Mouse
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6
Molecular Weight of Antigen 56kDa
Positive Control Tonsil or basal cell carcinoma
Purification Method Protein A/G Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286887
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.