missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 6 Antibody (SPM269), Novus Biologicals™

Mouse Monoclonal Antibody

£232.00 - £401.00

Specifications

Antigen Cytokeratin 6
Clone SPM269
Concentration 0.2 mg/mL
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18426111
View Documents
Novus Biologicals
NBP2-34358-0.02mg
0.02 mg
£232.00
0.02mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18460602
View Documents
Novus Biologicals
NBP2-34358-0.1mg
0.1 mg
£401.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytokeratin 6 Monoclonal antibody specifically detects Cytokeratin 6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

Cytokeratin 6
0.2 mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cell Biology, Cellular Markers, Stem Cell Markers
10 mM PBS with 0.05% BSA
Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6
A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human Cytokeratin 6
Primary
Store at 4C.
SPM269
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
Monoclonal
Purified
RUO
Human, Mouse
P02538, P04259, P48668
3853
IgG2a κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.