missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 5/8 Antibody (SPM268), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47823AF405
This item is not returnable.
View return policy
Description
Cytokeratin 5/8 Monoclonal antibody specifically detects Cytokeratin 5/8 in Human, Mouse, Rat, Porcine, Bovine, Canine, Hamster, Ferret, Sheep, Chicken (Negative), Xenopus (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| Cytokeratin 5/8 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5 | |
| Cytoskeletal preparation from HeLa cells | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3852 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM268 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction