missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 5/6 Antibody (AbM58019-7H4) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43564
This item is not returnable.
View return policy
Description
Cytokeratin 5/6 Monoclonal antibody specifically detects Cytokeratin 5/6 in Human samples. It is validated for ELISA,Immunohistochemistry
Specifications
| Cytokeratin 5/6 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry | |
| 58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 | |
| The original antibody was generated by immunizing 4-6 weeks old female ICR mice with recombinant protein comprising amino acids 488-590 of cytokeratin 5 in equal volume of Freund's incomplete adjuvant. (Uniprot# P13647) | |
| 0.2 mg | |
| Cell Biology, Cellular Markers | |
| 3852 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry | |
| AbM58019-7H4 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction