missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Cytokeratin 4 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16876224
|
Proteintech
16572-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16866224
|
Proteintech
16572-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in differentiated layers of the mucosal and esophageal epithelia with family member KRT13. Mutations in these genes have been associated with White Sponge Nevus, characterized by oral, esophageal, and anal leukoplakia. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.Specifications
| Cytokeratin 4 | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P19013 | |
| 3851 | |
| Cytokeratin 4 Fusion Protein Ag9854 | |
| Primary | |
| -20°C | |
| KRT4 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CK 4, CK4, CYK4, Cytokeratin 4, K4, keratin 4, KRT4, Type II keratin Kb4 | |
| KRT4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title