missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 4 Mouse anti-Human, Clone: KRT4/2804, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Cytokeratin 4 |
|---|---|
| Clone | KRT4/2804 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array |
Description
Cytokeratin 4 Monoclonal specifically detects Cytokeratin 4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Cytokeratin 4 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3851 | |
| IgG1 κ | |
| Protein A or G purified |
| KRT4/2804 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CK4, CK-4, CYK4cytokeratin-4, Cytokeratin-4, FLJ31692, K4cytokeratin 4, keratin 4, keratin, type II cytoskeletal 4, Keratin-4, Type-II keratin Kb4 | |
| Recombinant fragment (around aa 181-292) of human Cytokeratin 4 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title