missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 4 Antibody (KRT4/2804), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08583AF532
This item is not returnable.
View return policy
Description
Cytokeratin 4 Monoclonal antibody specifically detects Cytokeratin 4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Cytokeratin 4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CK4, CK-4, CYK4cytokeratin-4, Cytokeratin-4, FLJ31692, K4cytokeratin 4, keratin 4, keratin, type II cytoskeletal 4, Keratin-4, Type-II keratin Kb4 | |
| Recombinant fragment (around aa 181-292) of human Cytokeratin 4 protein (exact sequence is proprietary) (Uniprot: P19013) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 3851 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
| KRT4/2804 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction