missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 37/38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106801
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KRT37/38.
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. [provided by RefSeq, Jul 2008].
Specifications
| Cytokeratin 37/38 | |
| Polyclonal | |
| Unconjugated | |
| KRT37 | |
| HA7; HA8; hair keratin; hair keratin, type I Ha7; Hair keratin, type I Ha8; HHA7; HHA8; HKA7; HKA8; K37; K38; keratin 37; keratin 37, type I; keratin 38; keratin 38, type I; keratin, hair, acidic, 7; keratin, hair, acidic, 8; keratin, type I cuticular Ha7; Keratin, type I cuticular Ha8; keratin-37; Keratin-38; KRT hHa8; KRT37; KRT38; KRTHA7; KRTHA8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8687, 8688 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O76014, O76015 | |
| KRT37, KRT38 | |
| A synthesized peptide derived from human KRT37(Accession O76014), corresponding to amino acid residues Y355-R405. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction