missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 35 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568044
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
KRT35 is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
Specifications
| Cytokeratin 35 | |
| Polyclonal | |
| Unconjugated | |
| KRT35 | |
| HA5, Ha-5, KRTHA5, hHa5, HHa5 hair keratin type I intermediate filament, K35, hair keratin, type I Ha5, hard keratin, type I, 5, keratin, hair, acidic, 5, keratin, type I cuticular Ha5, keratin-35 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3886, 53617 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92764, Q497I4 | |
| KRT35 | |
| Synthesized peptide derived from human Cytokeratin 35. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction