missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytokeratin 34 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA567227

Product Code. 15979894

  • £463.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ATTNASGNSCG PCGTSQKGCC N Highest antigen sequence identity to the following orthologs - mouse 64%, rat 59%.

The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 34
Polyclonal
Unconjugated
Krt34
4733401E01Rik; Ha2; HA4; Ha-4; hair keratin; Hair keratin, type I Ha4; hard keratin, type I, 4; hHa4; HKA4; K34; keratin 34; keratin 34, type I; keratin complex 1, acidic, gene 4; keratin, hair, acidic, 4; keratin, type I cuticular Ha4; Keratin-34; KRT hHa4; Krt1-4; Krt34; KRTHA4
Rabbit
Antigen affinity chromatography
RUO
3885
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O76011
Krt34
Recombinant Human Cytokeratin 34. Recombinant protein control fragment (Product #RP-107865).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.