missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563083
This item is not returnable.
View return policy
Description
Immunogen sequence: ERDNAELENL IRERSQQQEP LLC Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 74%.
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
Specifications
| Cytokeratin 31 | |
| Polyclonal | |
| Unconjugated | |
| KRT31 | |
| Ha1; Ha-1; hair keratin; Hair keratin, type I Ha1; hard keratin, type I, 1; hHa1; HKA1; HKA-1; K31; Ka25; keratin 31; keratin 31, type I; keratin complex 1, acidic, gene 1; keratin complex 1, gene 1; keratin, hair, acidic,1; keratin, type I cuticular Ha1; Keratin-31; Kha1; KRT hHa1; Krt1-1; Krt31; KRTHA1; MKHA-1; type I hair keratin KA25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3881 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15323 | |
| KRT31 | |
| Recombinant protein corresponding to Human Cytokeratin 31. Recombinant protein control fragment (Product #RP-104497). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction