missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587646
This item is not returnable.
View return policy
Description
Immunogen sequence: WNLLQQQGTS SISGTNNLEP LFENHINYLR SYLDNILGER GRLDSELKNM EDLVEDFKKK YEDEINKRTA AENEFVTLKK DVDSAYMNKV ELQAKVDALI DEIDFLRTLY DAELSQMQSH ISDTSVVLSM DNNRSLDLDS IIAEVRAQYE DIAQRSKAEA E; Positive Samples: H460.
Cytokeratin 3 is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the corneal epithelium with family member KRT12 and mutations in these genes have been associated with Meesmann's Corneal Dystrophy. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. (NCBI Gene description).
Specifications
| Cytokeratin 3 | |
| Polyclonal | |
| Unconjugated | |
| KRT3 | |
| 65 kDa cytokeratin; CK3; CK-3; cytokeratin 3; cytokeratin type II, component Ib/c; cytokeratin-3; K3; K3 keratin; keratin 3; keratin 3, type II; keratin K3; keratin, type II cytoskeletal 3; Keratin-3; KERIB; KRT3; Type-II keratin Kb3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3850 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P12035 | |
| KRT3 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction