missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 3 Antibody (KRT3/2130) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79852
This item is not returnable.
View return policy
Description
Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Cytokeratin 3 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| KRT3/2130 | |
| Unconjugated | |
| 10 mM PBS with No Preservative | |
| KRT3 | |
| Recombinant full-length human Cytokeratin 3 protein (Uniprot: P12035) | |
| 100 μg | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction