missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 3/2p Antibody (2Q1040(AE5)), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34844AF700
This item is not returnable.
View return policy
Description
Cytokeratin 3/2p Monoclonal antibody specifically detects Cytokeratin 3/2p in Human, Mouse, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Cytokeratin 3/2p | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 65 KDa Cytokeratin, CK3, CK-3, cytokeratin-3, Keratin, keratin 3, Keratin, Type II Cytoskeletal 3, Keratin-3, Type II Cytoskeletal 3, Type-II keratin Kb3 | |
| Rabbit corneal epithelial keratin | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Bovine, Rabbit | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 2Q1040(AE5) | |
| 50mM Sodium Borate | |
| Mouse | |
| Ion exchange chromatography | |
| RUO | |
| 3850 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction