missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | Cytokeratin 27 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246352
|
Novus Biologicals
NBP2-55440 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637277
|
Novus Biologicals
NBP2-55440-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 27 Polyclonal specifically detects Cytokeratin 27 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 27 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 342574 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RRLGSCGGTGSVRLSSGGAGFGAGNTCGVPGIGSGFS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CK-27, Cytokeratin-27, K25C, K25IRS3, K27, Keratin 25C, Keratin 27, Keratin 27, Type I, Keratin, Type I Cytoskeletal 27, Keratin-25C, Keratin-27, KRT25C, Type I Inner Root Sheath Specific Keratin 25 Irs3, Type I Inner Root Sheath-Specific Keratin-K25irs3 | |
| KRT27 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title