missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 26 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82796-0.1ML
This item is not returnable.
View return policy
Description
Cytokeratin 26 Polyclonal specifically detects Cytokeratin 26 in Human samples. It is validated for Western Blot.
Specifications
| Cytokeratin 26 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 353288 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| CK26, CK-26, cytokeratin-26, K25B, K25IRS2, K26, Keratin 25B, Keratin, Type I Cytoskeletal 26, keratin-25B, Keratin-26, KRT25B, KRT26, Type I Inner Root Sheath Specific Keratin 25 Irs2, Type I Inner Root Sheath-Specific Keratin-K25irs2 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human Cytokeratin 26 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction