missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£470.00
Specifications
| Antigen | Cytokeratin 26 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Cytokeratin 26 Polyclonal specifically detects Cytokeratin 26 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 26 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 353288 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MSFRLSGGSRRICSRTGSGRLSGGGTGFVAGNVCVGSGARSSFSCTLEGISSGGSFCNSGGGLGSGACAGFLGNEHSLLSGNEKV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CK26, CK-26, cytokeratin-26, K25B, K25IRS2, K26, Keratin 25B, Keratin, Type I Cytoskeletal 26, keratin-25B, Keratin-26, KRT25B, KRT26, Type I Inner Root Sheath Specific Keratin 25 Irs2, Type I Inner Root Sheath-Specific Keratin-K25irs2 | |
| KRT26 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title