missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553524
This item is not returnable.
View return policy
Description
Immunogen sequence: KHHVPSDFNV NVKVDTGPRE DLIKVLEDMR QEYELIIKKK HRDLDTWYKE QSAAMSQEAA SPATVQSRQG DIHELKRTFQ ALEIDLQAQY STKSALENML SETQSRYSCK LQDMQEIISH YEEELTQLR Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
Specifications
| Cytokeratin 23 | |
| Polyclonal | |
| Unconjugated | |
| KRT23 | |
| C HAIK1; CK23; CK-23; cytokeratin 23; cytokeratin-23; DKFZP434G032; HAIK1; histone deacetylase inducible keratin 23; hyperacetylation-inducible type I keratin; K23; Ka23; keratin 23; keratin 23 (histone deacetylase inducible); keratin 23, type I; keratin complex 1, acidic, gene 23; keratin, type I cytoskeletal 23; Keratin-23; Krt1-23; KRT23; MGC26158; type I intermediate filament cytokeratin; type I keratin KA23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25984 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C075 | |
| KRT23 | |
| Recombinant protein corresponding to Human Cytokeratin 23. Recombinant protein control fragment (Product #RP-89653). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction