missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 23 Monoclonal Antibody (K56004_3E9), Invitrogen™
Mouse Monoclonal Antibody
£390.00 - £586.00
Specifications
| Antigen | Cytokeratin 23 |
|---|---|
| Clone | K56004_3E9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30206753
|
Invitrogen™
MA555746 |
50 μL |
£390.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30195459
|
Invitrogen™
MA556126 |
200 μL |
£586.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GTREESKSSM KVSATPKIKA
Cytokeratin 23 Monoclonal antibody specifically detects Cytokeratin 23 in Human samples. It is validated for Western BlotSpecifications
| Cytokeratin 23 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| C HAIK1; CK23; CK-23; cytokeratin 23; cytokeratin-23; DKFZP434G032; HAIK1; histone deacetylase inducible keratin 23; hyperacetylation-inducible type I keratin; K23; Ka23; keratin 23; keratin 23 (histone deacetylase inducible); keratin 23, type I; keratin complex 1, acidic, gene 23; keratin, type I cytoskeletal 23; Keratin-23; Krt1-23; KRT23; MGC26158; type I intermediate filament cytokeratin; type I keratin KA23 | |
| Human KRT23 peptide (immunogen range is 381-400). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| KRT23 |
| K56004_3E9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9C075 | |
| 25984 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title