Learn More
Cytokeratin 20 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-17329-100UL
Description
The Cytokeratin 20 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cytokeratin 20. This antibody recognizes human, mouse, rat, rabbit antigen. The Cytokeratin 20 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Cytokeratin 20 | |
| 1000 μg/mL | |
| CK 20, CK20, Cytokeratin 20, K20, keratin 20, KRT20, KRT21, Protein IT | |
| 100 μL | |
| 54474 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| 150 kDa | |
| IgG |