missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 20 Monoclonal Antibody (CL9390)

Description
Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 69%.
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
Specifications
Specifications
| Antigen | Cytokeratin 20 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL9390 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KRT20 |
| Gene Accession No. | P35900 |
| Gene Alias | 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?