Learn More
Cytokeratin 20 Antibody (CL9390) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88932R
Product Code. 30295933
  • £558.00 / 0.10mL
Description
Cytokeratin 20 Monoclonal antibody specifically detects Cytokeratin 20 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 20
Monoclonal
DyLight 550
CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT
This antibody was developed using a recombinant protein derived from P35900, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers
54474
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL9390
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified