missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Rabbit anti-Human, Mouse, Clone: KRT19/1959R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | Cytokeratin 19 |
|---|---|
| Clone | KRT19/1959R |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 19 | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Neuroscience, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3880 | |
| IgG | |
| Protein A or G purified |
| KRT19/1959R | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, MGC15366 | |
| Recombinant human full-length Cytokeratin 19 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title