missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytokeratin 19 Monoclonal Antibody (4E8)
GREENER_CHOICE

Product Code. 12857902
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12857902 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12857902 Supplier Invitrogen™ Supplier No. MA515884

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-15884 targets KRT19 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15884 immunogen is purified recombinant fragment of human KRT19 expressed in E. Coli. . MA5-15884 detects KRT19 which has a predicted molecular weight of approximately 41kDa.

Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 19
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4E8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene KRT19
Gene Accession No. P08727
Gene Alias 40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; Krt19; LOC100344434; MGC15366; Type I keratin Ka19
Gene Symbols KRT19
Host Species Mouse
Immunogen Purified recombinant fragment of human KRT19 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3880
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.