missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-687PE
This item is not returnable.
View return policy
Description
Cytokeratin 19 Polyclonal antibody specifically detects Cytokeratin 19 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| Cytokeratin 19 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| PE | |
| 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, MGC15366 | |
| Synthetic peptide derived from the C-terminal region of human Cytokeratin 19 (between residues 350-400) [UniProt P08727] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuroscience, Stem Cell Markers | |
| 3880 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction