missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£188.00 - £426.00
Specifications
| Antigen | Cytokeratin 19 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:1000, Simple Western 1:1000, Flow Cytometry 2-5 ug/million cells, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18045084
|
Novus Biologicals
NB100-687SS |
0.025 mL |
£188.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18728013
|
Novus Biologicals
NB100-687 |
£426.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Cytokeratin 19 Polyclonal specifically detects Cytokeratin 19 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 19 | |
| Western Blot 1:1000, Simple Western 1:1000, Flow Cytometry 2-5 ug/million cells, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:500 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Cytoskeleton Markers, Stem Cell Markers | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
| KRT19 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, and monkey. Rat reactivity reported in scientific literature (PMID: 25401473). |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P08727 | |
| 3880 | |
| Synthetic peptide derived from the C-terminal region of human Cytokeratin 19 (between residues 350-400) [UniProt P08727] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title