missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Antibody (963420) [DyLight 650], Novus Biologicals™

Product Code. 30735854 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735854 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735854 Supplier Novus Biologicals Supplier No. FAB35061W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytokeratin 19 Monoclonal antibody specifically detects Cytokeratin 19 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry,Dual RNAscope ISH-IHC
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 19
Applications Immunohistochemistry, Western Blot, Immunocytochemistry, Dual RNAScope ISH-IHC
Classification Monoclonal
Clone 963420
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, MGC15366
Host Species Mouse
Immunogen E.coli-derived recombinant human Cytokeratin 19, Gln311-Lys370, Accession # P08727
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3880
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.