Learn More
Cytokeratin 18 Rabbit, anti-Human, Clone: 4F2, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82817-5-RR-20UL
Description
The Cytokeratin 18 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Cytokeratin 18. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 18 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Cytokeratin 18 | |
| 4F2 | |
| CK 18, CYK18, Cytokeratin 18, K18, keratin 18, KRT18 | |
| 20 μL | |
| 3875 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |