missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 18 Monoclonal Antibody (DC-10), Biotin

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA119476
This item is not returnable.
View return policy
Description
This antibody reacts with cytokeratin 18 (45 kDa). Cytokeratins are members of intermediate filaments subfamily of intracellular proteins, represented in epithelial tissues.
Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P05783 | |
| KRT18 | |
| Human breast carcinoma cell line PMC-42. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| DC-10 | |
| Biotin | |
| KRT18 | |
| cb83; Cell proliferation-inducing gene 46 protein; CK 18; CK18; CK-18; CYK18; cytokeratin 18; cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin18; cytokeratin-18; dapk1; dorsal aorta proneprin kinesin-1; DreK18; Endo B; I79_023185; K 18; K18; keratin 18; keratin 18, type I; keratin 18a, tandem duplicate 1; keratin complex 1, acidic, gene 18; keratin D; Keratin, type I cytoskeletal 18; Keratin18; keratin-18; Kerd; KRT 18; Krt1-18; KRT18; krt18 {ECO:0000250; krt18a.1; LOC100008885; PIG46; sb:cb83; type I cytoskeletal 18; UniProtKB:P05783}; wu:fa13f02; wu:fb36b04 | |
| Mouse | |
| Size-exclusion Chromatography | |
| RUO | |
| 3875 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction