missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (LDK18), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50463PE
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Mouse, Porcine samples. It is validated for Flow Cytometry.
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| KRT18 | |
| Synthetic peptide of human Cytokeratin 18 (CSETNDTKVLRH-COOH) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers | |
| 3875 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| LDK18 | |
| Flow Cytometry | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction