Learn More
Cytokeratin 18 Antibody (DC10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33082APC
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Bovine (Negative), Canine (Negative), Hamster (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative), Sheep (Negative) samples. It is validated for Flow Cytometry.
Specifications
| Flow Cytometry | |
| 45 kDa | |
| Bovine (Negative), Canine (Negative) | |
| IgG1 κ |
| KRT18 | |
| Protein A or G purified | |
| Purified |