missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 18 Antibody (C-04 (same as Ks18.04)) - Azide and BSA Free, Novus Biologicals™

Product Code. 18393473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393473 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393473 Supplier Novus Biologicals Supplier No. NBP311485

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline, Goat, Hamster, Ferret, Monkey, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 18
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone C-04 (same as Ks18.04)
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml
Formulation 10 mM PBS
Gene Alias Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18
Host Species Mouse
Immunogen Cytoskeletal preparation extracted from the human vulvar squamous carcinoma cell line A431
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3875
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline, Goat, Hamster, Ferret, Monkey, Sheep, Chicken (Negative), Xenopus (Negative)
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.