missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Cytokeratin 17 |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16811775
|
Proteintech
22230-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16801775
|
Proteintech
22230-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes the type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in this gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex.Specifications
| Cytokeratin 17 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q04695, Q9QWL7 | |
| 16667, 3872 | |
| Cytokeratin 17 Fusion Protein Ag17575 | |
| Primary | |
| -20°C | |
| KRT17 |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 39.1, CK 17, Cytokeratin 17, K17, keratin 17, KRT17, PC, PC2, PCHC1 | |
| Krt17 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title