missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Mouse anti-Human, Clone: KRT17/4604, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Cytokeratin 17 |
|---|---|
| Clone | KRT17/4604 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
Cytokeratin 17 Monoclonal specifically detects Cytokeratin 17 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 17 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 3872 | |
| IgG1 κ | |
| Protein A or G purified |
| KRT17/4604 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
| Recombinant full-length human Cytokeratin 17 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title