missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 17 Recombinant Rabbit Monoclonal Antibody (9O6X9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535401
This item is not returnable.
View return policy
Description
Immunogen sequence: MTTSIRQFTS SSSIKGSSGL GGGSSRTSCR LSGGLGAGSC RLGSAGGLGS TLGGSSYSSC YSFGSGGGYG SSFGGVDGLL AGGEKATMQN LNDRLASYLD.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9 - 7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Specifications
| Cytokeratin 17 | |
| Recombinant Monoclonal | |
| 0.53 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q04695, Q6IFU8, Q9QWL7 | |
| KRT17 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cytokeratin 17 (KRT17) (Q04695). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9O6X9 | |
| Unconjugated | |
| KRT17 | |
| 39.1; CK17; CK-17; cytokeratin-17; hard keratin; K17; Ka17; keratin 17; keratin 17, type I; keratin complex 1, acidic, gene 17; keratin, type I cytoskeletal 17; Keratin-17; Krt1-17; Krt17; krt17 {ECO:0000250; PC; PC2; PCHC1; Type I keratin Ka17; UniProtKB:Q04695} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16667, 287702, 3872 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction