missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Antibody (4D2) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43565
This item is not returnable.
View return policy
Description
Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human samples. It is validated for ELISA,Immunohistochemistry
Specifications
| Cytokeratin 17 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry | |
| 39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
| The original antibody was generated by immunizing female BALB/c mice with an immunogenic polypeptide containing amino acids 410-432 (TIVEEVQDGKVISSREQVHQTTR) of human cytokeratin 17 conjugated with KLH. (Uniprot# Q04695) | |
| 0.2 mg | |
| Cancer | |
| 3872 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry | |
| 4D2 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction