missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 17/19 Antibody (1080201) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30762732 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762732 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762732 Supplier Novus Biologicals Supplier No. FAB11568MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytokeratin 17/19 Monoclonal antibody specifically detects Cytokeratin 17/19 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 17/19
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1080201
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias 39.1, 40-kDa keratin intermediate filament, CK-17, CK19, CK-19, cytokeratin 19, cytokeratin-17, K17, K19, K1CS, keratin 19, type I, keratin, type I cytoskeletal 17, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, KRT19, PC, PC2, PCHC1
Host Species Mouse
Immunogen Cytokeratin 17/19 containing synthetic peptide, Accession # Q04695
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3872
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.