missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 16 Rabbit anti-Human, Mouse, Rat, Clone: 8L6R4, Novus Biologicals™

Product Code. 18328471
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328471 20 μg 20µL
18365286 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18328471 Supplier Bio-Techne Supplier No. NBP31667820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytokeratin 16 Monoclonal antibody specifically detects Cytokeratin 16 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 16
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8L6R4
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cytokeratin 16 (P08779). MTTCSRQFTSSSSMKGSCGIgGGIgGGSSRISSVLAGGSCRAPSTYGGGLSVSSRFSSGGACGLGGGYGGGFSSSSSFGSGFGGGYGGGLGAGFGGGLGA
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3868
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.