missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 16 Mouse anti-Human, Clone: SPM264, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00
Specifications
| Antigen | Cytokeratin 16 |
|---|---|
| Clone | SPM264 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1 to 2 μg/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
Cytokeratin 16 Monoclonal specifically detects Cytokeratin 16 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Cytokeratin 16 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3868 | |
| IgG1 κ | |
| Protein A or G purified |
| SPM264 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK | |
| Recombinant fragment from the C-terminal of human Cytokeratin 16 | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title