missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 16 Monoclonal Antibody (LL025), Invitrogen™

Product Code. 11394903
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394903 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11394903 Supplier Invitrogen Supplier No. MA513730

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-13730 targets Cytokeratin 16 in immunofluorescence and immunohistochemistry (paraffin) applications and shows reactivity with Human samples. Staining of formalin/paraffin tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6.0 for 10-20 minutes followed by cooling at room temperature for 20 minutes. The MA5-13730 immunogen is a synthetic peptide from the C-terminus of human cytokeratin 16.

Cytokeratin 16 Monoclonal antibody specifically detects Cytokeratin 16 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LL025
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene KRT16
Gene Accession No. P08779
Gene Alias AI324768; CK16; CK-16; cytokeratin 16; cytokeratin-16; FNEPPK; focal non-epidermolytic palmoplantar keratoderma; K16; K1CP; Ka16; keratin 16; keratin 16, type I; keratin complex 1, acidic, gene 16; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin, type I cytoskeletal 16; Keratin-16; Krt1-16; Krt16; KRT16A; NEPPK; PC1; type I keratin KA16
Gene Symbols KRT16
Host Species Mouse
Immunogen A synthetic peptide from the C-terminus of human cytokeratin 16
Purification Method Ammonium sulfate precipitation
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3868
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.