missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytokeratin 15 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA582854

Product Code. 16339814

  • £502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RLEQEIATYR SLLEGQDAKM AGIGIREASS GGGGSSSNFH INVEESVDGQ VVSSHKREI Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 80%.

Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21. 2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 15
Polyclonal
Unconjugated
KRT15
AI528832; C K1CO; CK15; CK-15; cytokeratin 15; cytokeratin-15; K15; K1CO; Ka15; keratin 15; keratin 15, type I; keratin complex 1, acidic, gene 15; keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; Krt1-15; KRT15; krt15 {ECO:0000250; KRTB; type I cytoskeletal 15; type I keratin KA15; UniProtKB:P19012}
Rabbit
Antigen affinity chromatography
RUO
3866
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P19012
KRT15
Recombinant protein corresponding to Human KRT15. Recombinant protein control fragment (Product #RP-93406).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.